3-tert-Butylbenzenesulfonyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: 3-(tert-Butyl)benzene-1-sulphonyl chloride

A127762-100MG A127762-1G A127762-250MG
76688-516EA 33.95 USD
76688-516 77624-066 77624-068
3-tert-Butylbenzenesulfonyl chloride 97%
3-tert-Butylbenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClO₂S
MW: 232.73 g/mol
MDL Number: MFCD03789234
CAS Number: 2905-26-2
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR