Sodium benzenesulfinate 95%

Supplier: Ambeed
Warning

A127123-1000G A127123-100G A127123-10G A127123-25G A127123-500G
76688-484EA 52 USD
76688-484 76688-486 76688-488 76688-490 76688-492
Sodium benzenesulfinate 95%
Sodium benzenesulphinate

Precaución: for Research Use Only

Formula: C₆H₅NaO₂S
MDL Number: MFCD00013135
CAS Number: 873-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR