7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde, 2-Bromo-3,4-ethylenedioxythiophene-5-carboxaldehyde

A127115-1G A127115-250MG A127115-5G
76688-478EA 68.96 USD
76688-478 76688-480 76688-482
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde 95%
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde

Precaución: for Research Use Only

Formula: C₇H₅BrO₃S
MW: 249.08 g/mol
MDL Number: MFCD08690306
CAS Number: 852054-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR