Biotin hydrazide 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide
A126955-100MG
A126955-1G
A126955-1MG
A126955-250MG
A126955-25MG
A126955-5G
76688-456EA
51
USD
76688-456
76688-458
76688-460
76688-462
76688-464
76688-466
Biotin hydrazide 95%
Biotin hydrazide
Precaución: for Research Use Only
Formula:
C₁₀H₁₈N₄O₂S |
MDL Number:
MFCD00078532 CAS Number: 66640-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...