Scutellarin 97%

Supplier: Ambeed
Warning

Synonyms: (2S,3S,4S,5R,6S)-6-((5,6-Dihydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-chromen-7-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid

A126805-25MG A126805-50MG A126805-1G A126805-250MG A126805-5G
76688-450EA 5.31 USD
76688-450 76688-452 77599-834 77599-836 77599-838
Scutellarin 97%
Scutellarin

Precaución: for Research Use Only

Formula: C₂₁H₁₈O₁₂
MW: 462.36 g/mol
MDL Number: MFCD01861503
CAS Number: 27740-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR