Bis(2-ethylhexyl)maleate 98%

Supplier: Ambeed
Warning

Synonyms: Maleic Acid Di(2-ethylhexyl) Ester, Maleic Acid Dioctyl Ester, Bis(2-ethylhexyl) maleate, Di(2-ethylhexyl) Maleate

A126729-100G A126729-25G A126729-500G
76688-444EA 26 USD
76688-444 76688-446 76688-448
Bis(2-ethylhexyl)maleate 98%
Bis(2-ethylhexyl)maleate

Precaución: for Research Use Only

Formula: C₂₀H₃₆O₄
MW: 340.50 g/mol
MDL Number: MFCD00027250
CAS Number: 142-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR