(R)-2-Amino-2-phenylacetic acid hydrochloride 98%

Supplier: Ambeed
Warning

A126674-100G A126674-1G A126674-25G A126674-5G
76688-436EA 294.94 USD
76688-436 76688-438 76688-440 76688-442
(R)-2-Amino-2-phenylacetic acid hydrochloride 98%
(R)-2-Amino-2-phenylacetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO₂
MW: 187.62 g/mol
MDL Number: MFCD08443602
CAS Number: 25705-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR