2-Isobutyl-4,5-dimethyl-3-thiazoline (mixture of isomers) 95% mix

Supplier: Ambeed
Warning

Synonyms: 2-Isobutyl-4,5-dimethyl-3-thiazoline, 4,5-Dimethyl-2-isobutyl-3-thiazoline

A126617-100G A126617-25G A126617-5G
76688-426EA 112.46 USD
76688-426 76688-428 76688-430
2-Isobutyl-4,5-dimethyl-3-thiazoline (mixture of isomers) 95% mix
2-Isobutyl-4,5-dimethyl-3-thiazoline (mixture of isomers)

Precaución: for Research Use Only

Formula: C₉H₁₇NS
MW: 171.30 g/mol
MDL Number: MFCD01680822
CAS Number: 65894-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR