2-Propoxybenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-n-Propoxybenzoic acid

A126543-100G A126543-10G A126543-500G A126543-5G A126543-25G
76688-418EA 42.44 USD
76688-418 76688-420 76688-422 76688-424 77313-050
2-Propoxybenzoic acid 97%
2-Propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
MDL Number: MFCD01075687
CAS Number: 2100-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR