Amikacin disulphate 98% (contains ~10% H₂O)

Supplier: Ambeed
Warning

Synonyms: (S)-4-Amino-N-((1R,2S,3S,4R,5S)-5-amino-2-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4-(((2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-3-hydroxycyclohexyl)-2-hydroxybutanamide bis(sulphate)

A125426-100G A125426-10G A125426-1G A125426-25G A125426-5G
76688-362EA 177.18 USD
76688-362 76688-364 76688-366 76688-368 76688-370
Amikacin disulphate 98% (contains ~10% H₂O)
Amikacin disulphate

Precaución: for Research Use Only

Formula: C₂₂H₄₃N₅O₁₃·2H₂SO₄
MW: 781.77 g/mol
MDL Number: MFCD00167475
CAS Number: 39831-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR