Butenafine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-(tert-Butyl)benzyl)-N-methyl-1-(naphthalen-1-yl)methanamine hydrochloride, Butenafine HCl

A125188-10G A125188-1G A125188-1MG A125188-25G A125188-5G A125188-100G
76688-342EA 38.2 USD
76688-342 76688-344 76688-346 76688-348 76688-350 77599-812
Butenafine hydrochloride 98%
Butenafine hydrochloride

Precaución: for Research Use Only

Formula: C₂₃H₂₇N
MW: 353.93 g/mol
MDL Number: MFCD00917064
CAS Number: 101827-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR