Methyl-N-acetyl-L-valinate 95%

Supplier: Ambeed
Warning

Synonyms: (S)-Methyl 2-acetamido-3-methylbutanoate, Ac-Val-OMe

A124890-250MG A124890-5G A124890-1G
76688-324EA 22 USD
76688-324 76688-326 77876-904
Methyl-N-acetyl-L-valinate 95%
Methyl-N-acetyl-L-valinate

Precaución: for Research Use Only

Formula: C₈H₁₅NO₃
MW: 173.212 g/mol
MDL Number: MFCD00080847
CAS Number: 1492-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR