Adamantane 97%

Supplier: Ambeed
Warning

Synonyms: Tricyclo[3.3.1.1(3,7)]decane

A124873-100G A124873-25G A124873-500G A124873-1000G
76688-318EA 9.55 USD
76688-318 76688-320 76688-322 77599-808
Adamantane 97%
Adamantane

Precaución: for Research Use Only

Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00074719
CAS Number: 281-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR