5-Nitrooxindole 98%

Supplier: Ambeed
Warning

Synonyms: 5-Nitroindolin-2-one, 5-Nitro-2-oxindole

A124868-10G A124868-1G A124868-25G A124868-5G
76688-310EA 66.84 USD
76688-310 76688-312 76688-314 76688-316
5-Nitrooxindole 98%
5-Nitrooxindole

Precaución: for Research Use Only

Formula: C₈H₆N₂O₃
MW: 178.15 g/mol
MDL Number: MFCD00456999
CAS Number: 20870-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR