(R)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate 97%

Supplier: Ambeed
Warning

Synonyms: 4-[4-(Hexyloxy)benzoyloxy]benzoic acid (R)-2-Octyl ester, (R)-Octan-2-yl 4-((4-(hexyloxy)benzoyl)oxy)benzoate

A124771-100MG A124771-1G A124771-250MG A124771-5G A124771-25G
76688-302EA 12.74 USD
76688-302 76688-304 76688-306 76688-308 77566-404
(R)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate 97%
(R)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate

Precaución: For research use only.

Formula: C₂₈H₃₈O₅
MW: 454.61 g/mol
MDL Number: MFCD21603881
CAS Number: 133676-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR