D-(+)-Raffinose pentahydrate 98%

Supplier: Ambeed
Warning

Synonyms: D-(+)-Raffinose pentahydrate, Gossypose pentahydrate, (3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-((((3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol pentahydrate, Melitriose pentahydrate, O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside, Melitose pentahydrate

A124248-100G A124248-10G A124248-250MG A124248-25G A124248-500G A124248-5G A124248-1000G
76688-266EA 47 USD
76688-266 76688-268 76688-270 76688-272 76688-274 76688-276 77878-911
D-(+)-Raffinose pentahydrate 98%
D-(+)-Raffinose pentahydrate

Precaución: for Research Use Only

Formula: C₁₈H₃₂O₁₆·5H₂O
MW: 594.52 g/mol
Melting Pt: 80
MDL Number: MFCD00071590
CAS Number: 17629-30-0
Merck Index: 12,08279

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR