(S)-tert-Butyl-5-azaspiro[2.4]heptan-7-ylcarbamate 98%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl 5-azaspiro[2.4]heptan-7-ylcarbamate

A124184-100MG A124184-250MG A124184-5G A124184-1G
76688-260EA 32.89 USD
76688-260 76688-262 76688-264 77624-040
(S)-tert-Butyl-5-azaspiro[2.4]heptan-7-ylcarbamate 98%
(S)-tert-Butyl-5-azaspiro[2.4]heptan-7-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂
MW: 212.29 g/mol
MDL Number: MFCD09955395
CAS Number: 127199-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR