4'-(Difluoromethoxy)acetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-(Difluoromethoxy)phenyl)ethanone

A123896-100G A123896-10G A123896-1G A123896-25G A123896-5G
76688-234EA 741.57 USD
76688-234 76688-236 76688-238 76688-240 76688-242
4'-(Difluoromethoxy)acetophenone 97%
4'-(Difluoromethoxy)acetophenone

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.1600 g/mol
MDL Number: MFCD00042250
CAS Number: 83882-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR