Dibenzosuberenol 98%

Supplier: Ambeed
Warning

Synonyms: 1-Hydroxydibenzo[b,f]cycloheptene, 5H-Dibenzo[a,d][7]annulen-5-ol, 5-Dibenzosuberenol, Dibenzo[b,f]cyclohepten-1-ol, 1-Dibenzo[b,f]cycloheptenol

A122825-1G A122825-5G
76688-200EA 45.62 USD
76688-200 76688-202
Dibenzosuberenol 98%
Dibenzosuberenol

Precaución: for Research Use Only

Formula: C₁₅H₁₂O
MW: 208.26 g/mol
MDL Number: MFCD00003586
CAS Number: 10354-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR