Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, Ethyl 4-chloro-2-(methylsulphanyl)pyrimidine-5-carboxylate, Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate
A121862-100G
A121862-10G
A121862-25G
A121862-5G
76688-158EA
44.56
USD
76688-158
76688-160
76688-162
76688-164
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate 97%
Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate
Precaución: for Research Use Only
Formula:
C₈H₉ClN₂O₂S MW: 232.69 g/mol |
MDL Number:
MFCD00006085 CAS Number: 5909-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...