Ac-Glu(OtBu)-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-Acetyl-L-glutamic acid 5-tert-butyl ester

A120632-1G A120632-5G A120632-25G
76688-102EA 16 USD
76688-102 76688-104 77549-418
Ac-Glu(OtBu)-OH 98%
Ac-Glu(OtBu)-OH

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₅
MDL Number: MFCD00236754
CAS Number: 84192-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR