Cyclohexylamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Cyclohexylamine Hydrochloride

A120346-100G A120346-25G
76688-100EA 27 USD
76688-100 77623-998
Cyclohexylamine hydrochloride 98%
Cyclohexylamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃N
MW: 135.64 g/mol
MDL Number: MFCD00050540
CAS Number: 4998-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR