1-Bromo-4-ethylbenzene 97%

Supplier: Ambeed
Warning

A120307-1000G A120307-100G A120307-10G A120307-25G A120307-500G A120307-5G
76688-088EA 125 USD
76688-088 76688-090 76688-092 76688-094 76688-096 76688-098
1-Bromo-4-ethylbenzene 97%
1-Bromo-4-ethylbenzene

Precaución: for Research Use Only

Formula: C₈H₉Br
MDL Number: MFCD00000111
CAS Number: 1585-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR