Ac-Lys(Fmoc)-OH 96%

Supplier: Ambeed
Warning

A120304-100MG A120304-1G A120304-250MG A120304-5G A120304-25G
76688-082EA 5 USD
76688-082 76688-084 76688-086 77571-216 77876-874
Ac-Lys(Fmoc)-OH 96%
Ac-Lys(Fmoc)-OH

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₅
MDL Number: MFCD08458546
CAS Number: 148101-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR