7-Quinolinecarboxylic acid 98%

Supplier: Ambeed
Warning

A120178-100MG A120178-10G A120178-1G A120178-250MG A120178-5G
76688-066EA 12 USD
76688-066 76688-068 76688-070 76688-072 77599-728
7-Quinolinecarboxylic acid 98%
7-Quinolinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MDL Number: MFCD00047617
CAS Number: 1078-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR