Methyl-1-aminocyclobutanecarboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A119267-10G A119267-1G A119267-250MG A119267-5G A119267-25G
76688-042EA 6.37 USD
76688-042 76688-044 76688-046 76688-048 76688-050 77878-888
Methyl-1-aminocyclobutanecarboxylate hydrochloride 97%
Methyl-1-aminocyclobutanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₂
MDL Number: MFCD04115288
CAS Number: 92398-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR