Benzyl-DL-mandelate 98%

Supplier: Ambeed
Warning

Synonyms: DL-Mandelic Acid Benzyl Ester, Benzyl 2-hydroxy-2-phenylacetate

A119080-100G A119080-25G A119080-5G
76688-034EA 88.06 USD
76688-034 76688-036 76688-038
Benzyl-DL-mandelate 98%
Benzyl-DL-mandelate

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
MDL Number: MFCD00021856
CAS Number: 890-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR