2-Fluorocinnamic acid 98,5% predominantly trans

Supplier: Ambeed
Warning

Synonyms: 3-(2-Fluorophenyl)propenoic acid

A118916-100G A118916-5G A118916-25G
76688-030EA 127.31 USD
76688-030 76688-032 77599-716
2-Fluorocinnamic acid 98,5% predominantly trans
2-Fluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₂
MW: 166.15 g/mol
MDL Number: MFCD00004370
CAS Number: 451-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR