N-(tert-Butoxycarbonyl)-L-methionine 98%

Supplier: Ambeed
Warning

A118578-100G A118578-25G A118578-500G A118578-5G A118578-10G
76687-998EA 72 USD
76687-998 76688-000 76688-002 76688-004 77623-990
N-(tert-Butoxycarbonyl)-L-methionine 98%
N-(tert-Butoxycarbonyl)-L-methionine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄S
MDL Number: MFCD00065586
CAS Number: 2488-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR