Phenylpropiolic acid 95%

Supplier: Ambeed
Warning

Synonyms: Phenylacetylenecarboxylic acid, Phenylpropynoic acid, 3-Phenylpropiolic acid

A118568-100G A118568-10G A118568-1G A118568-25G A118568-500G A118568-5G
76687-986EA 248.26 USD
76687-986 76687-988 76687-990 76687-992 76687-994 76687-996
Phenylpropiolic acid 95%
Phenylpropiolic acid

Precaución: for Research Use Only

Formula: C₉H₆O₂
MW: 146.15 g/mol
MDL Number: MFCD00004361
CAS Number: 637-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR