N-(2-Cyanoethyl)-N-methylaniline 97%

Supplier: Ambeed
Warning

A117935-100G A117935-10G A117935-25G A117935-500G A117935-5G
76687-974EA 11 USD
76687-974 76687-976 76687-978 76687-980 77876-863
N-(2-Cyanoethyl)-N-methylaniline 97%
N-(2-Cyanoethyl)-N-methylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂
MDL Number: MFCD00019856
CAS Number: 94-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR