1-(Quinolin-8-yl)ethanone 95%

Supplier: Ambeed
Warning

A117921-100MG A117921-250MG A117921-1G
76687-970EA 57.29 USD
76687-970 76687-972 77566-160
1-(Quinolin-8-yl)ethanone 95%
1-(Quinolin-8-yl)ethanone

Precaución: For research use only.

Formula: C₁₁H₉NO
MW: 171.195 g/mol
MDL Number: MFCD00047620
CAS Number: 56234-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR