(±)-β-Citronellol 95+%

Supplier: Ambeed
Warning

Synonyms: DL-Citronellol, (±)-Citronellol, Citronellol, β-Citronellol, 3,7-Dimethyloct-6-en-1-ol

A117463-1000G A117463-25G A117463-500G A117463-100G
76687-944EA 66.84 USD
76687-944 76687-946 77549-356 77566-138
(±)-β-Citronellol 95+%
(±)-β-Citronellol

Precaución: For research use only.

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
MDL Number: MFCD00002935
CAS Number: 106-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR