trans-2-Phenylcyclopropanecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: trans-2-Phenylcyclopropane-1-carboxylic acid

A117369-100G A117369-1G A117369-250MG A117369-25G A117369-5G
76687-934EA 599.41 USD
76687-934 76687-936 76687-938 76687-940 76687-942
trans-2-Phenylcyclopropanecarboxylic acid 97%
trans-2-Phenylcyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MW: 162.1852 g/mol
MDL Number: MFCD00001292
CAS Number: 939-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR