2-(1H-Benzo[d][1,2,3]triazol-1-yl)-2-(((benzyloxy)carbonyl)amino)acetic acid 95%
Supplier: Ambeed
|
Warning
|
A116861-100MG
A116861-1G
A116861-250MG
A116861-5G
76687-906EA
14.86
USD
76687-906
76687-908
76687-910
76687-912
2-(1H-Benzo[d][1,2,3]triazol-1-yl)-2-(((benzyloxy)carbonyl)amino)acetic acid 95%
2-(1H-Benzo[d][1,2,3]triazol-1-yl)-2-(((benzyloxy)carbonyl)amino)acetic acid
Precaución: for Research Use Only
Formula:
C₁₆H₁₄N₄O₄ MW: 326.31 g/mol |
MDL Number:
MFCD00961511 CAS Number: 124676-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...