8-(4-Amino-1-methylbutylamino)-6-methoxyquinoline 95% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: Primaquine

A116308-1G A116308-250MG
76687-880EA 177.18 USD
76687-880 76687-882
8-(4-Amino-1-methylbutylamino)-6-methoxyquinoline 95% (by HPLC)
8-(4-Amino-1-methylbutylamino)-6-methoxyquinoline

Precaución: for Research Use Only

Formula: C₁₅H₂₁N₃O
MW: 259.3467 g/mol
MDL Number: MFCD00598906
CAS Number: 90-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR