Ethyl-7-bromoheptanoate 97%

Supplier: Ambeed
Warning

Synonyms: 7-Bromoheptanoic Acid Ethyl Ester, Ethyl 7-bromoheptanoate

A116102-10G A116102-1G A116102-25G A116102-5G A116102-100G
76687-862EA 25 USD
76687-862 76687-864 76687-866 76687-868 77549-310
Ethyl-7-bromoheptanoate 97%
Ethyl-7-bromoheptanoate

Precaución: for Research Use Only

Formula: C₉H₁₇BrO₂
MDL Number: MFCD00037103
CAS Number: 29823-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR