7-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine 96%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-3,4-dihydro-2H-1,4-benzoxazine

A116061-100MG A116061-10G A116061-1G A116061-250MG A116061-25G A116061-5G
76687-844EA 31.83 USD
76687-844 76687-846 76687-848 76687-850 76687-852 76687-854
7-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine 96%
7-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine

Precaución: for Research Use Only

Formula: C₈H₈BrNO
MW: 214.06 g/mol
MDL Number: MFCD09056750
CAS Number: 105679-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR