8-Methoxyquinoline 97%

Supplier: Ambeed
Warning

A115812-1G A115812-250MG A115812-25G A115812-5G
76687-800EA 12.74 USD
76687-800 76687-802 76687-804 76687-806
8-Methoxyquinoline 97%
8-Methoxyquinoline

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.19 g/mol
MDL Number: MFCD00957068
CAS Number: 938-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR