meso-tert-Butyl hexahydropyrrolo[3,4-c]pyrrole-2(1H)-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: cis-3,7-Diazabicyclo[3.3.0]octane, N3-BOC protected, cis-2-Boc-Hexahydropyrrol[3,4-c]pyrrole

A115706-100MG A115706-10G A115706-1G A115706-250MG A115706-25G A115706-5G
76687-780EA 10.61 USD
76687-780 76687-782 76687-784 76687-786 76687-788 76687-790
meso-tert-Butyl hexahydropyrrolo[3,4-c]pyrrole-2(1H)-carboxylate 97%
meso-tert-Butyl hexahydropyrrolo[3,4-c]pyrrole-2(1H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂
MW: 212.29 g/mol
MDL Number: MFCD04116224
CAS Number: 250275-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR