7-Bromobenzofuran-3(2H)-one 98%

Supplier: Ambeed
Warning

A115622-100MG A115622-1G A115622-250MG
76687-774EA 55.17 USD
76687-774 76687-776 76687-778
7-Bromobenzofuran-3(2H)-one 98%
7-Bromobenzofuran-3(2H)-one

Precaución: for Research Use Only

Formula: C₈H₅BrO₂
MW: 213.0300 g/mol
MDL Number: MFCD03160589
CAS Number: 519018-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR