2,2'-Dinaphthylamine 98%

Supplier: Ambeed
Warning

A115413-1G A115413-250MG A115413-25G A115413-5G
76687-766EA 21 USD
76687-766 76687-768 76687-770 76687-772
2,2'-Dinaphthylamine 98%
2,2'-Dinaphthylamine

Precaución: for Research Use Only

Formula: C₂₀H₁₅N
MDL Number: MFCD03093264
CAS Number: 532-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR