3,5-Bis(trifluoromethyl)benzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (3,5-Bis(trifluoromethyl)phenyl)methanol

A114974-100G A114974-1G A114974-250MG A114974-25G A114974-500G A114974-5G
76687-754EA 68.96 USD
76687-754 76687-756 76687-758 76687-760 76687-762 76687-764
3,5-Bis(trifluoromethyl)benzyl alcohol 98%
3,5-Bis(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₆F₆O
MW: 244.14 g/mol
MDL Number: MFCD00009907
CAS Number: 32707-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR