2-(2-(4-Benzhydrylpiperazin-1-yl)ethoxy)ethanol 98%

Supplier: Ambeed
Warning

Synonyms: Decloxizine

A114849-1G A114849-100MG A114849-250MG
76687-752EA 743 USD
76687-752 77878-861 77878-862
2-(2-(4-Benzhydrylpiperazin-1-yl)ethoxy)ethanol 98%
2-(2-(4-Benzhydrylpiperazin-1-yl)ethoxy)ethanol

Precaución: for Research Use Only

Formula: C₂₁H₂₈N₂O₂
MDL Number: MFCD00869150
CAS Number: 3733-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR