6-Nitro-2,3-dihydro-1H-isoindol-1-one 96%

Supplier: Ambeed
Warning

Synonyms: 6-Nitroisoindolin-1-one

A114682-100MG A114682-1G A114682-250MG A114682-5G
76687-734EA 12.74 USD
76687-734 76687-736 76687-738 76687-740
6-Nitro-2,3-dihydro-1H-isoindol-1-one 96%
6-Nitro-2,3-dihydro-1H-isoindol-1-one

Precaución: for Research Use Only

Formula: C₈H₆N₂O₃
MW: 178.145 g/mol
MDL Number: MFCD03093931
CAS Number: 110568-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR