Methyl 2-(((2'-cyano-[1,1'-biphenyl]-4-yl)methyl)amino)-3-nitrobenzoate 97%

Supplier: Ambeed
Warning

A113791-250MG A113791-25G A113791-1G A113791-5G
76687-702EA 6.37 USD
76687-702 77592-208 77623-944 77623-946
Methyl 2-(((2'-cyano-[1,1'-biphenyl]-4-yl)methyl)amino)-3-nitrobenzoate 97%
Methyl 2-(((2'-cyano-[1,1'-biphenyl]-4-yl)methyl)amino)-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₂₂H₁₇N₃O₄
MW: 387.38 g/mol
MDL Number: MFCD09029084
CAS Number: 139481-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR