Butyl laurate 99%

Supplier: Ambeed
Warning

Synonyms: Butyl dodecanoate, Dodecylic acid butyl ester, Dodecanoic acid butyl ester, Lauric acid butyl ester, n-Butyl dodecanoate

A113636-100G A113636-25G A113636-500G
76687-696EA 24.41 USD
76687-696 76687-698 76687-700
Butyl laurate 99%
Butyl laurate

Precaución: for Research Use Only

Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
MDL Number: MFCD00042870
CAS Number: 106-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR