8-Quinolinol sulphate 97%

Supplier: Ambeed
Danger

Synonyms: 8-Hydroxyquinoline sulphate, 8-Quinolinone sulphate, 8-Hydroxyquinolinium sulphate, 8-Hydroxyquinoline hemisulphate, Oxine sulphate, 8-Quinolinol sulphate, Quinolin-8-ol sulphate

A113110-25G A113110-500G A113110-100G
76687-674EA 7 USD
76687-674 76687-676 77623-936
8-Quinolinol sulphate 97%
8-Quinolinol sulfate

Precaución: for Research Use Only

Formula: C₉H₉NO₅S
MDL Number: MFCD00013095
CAS Number: 134-31-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR