Ethyl-3-methyl-4-nitrobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-methyl-4-nitrobenzoate

A113067-100G A113067-10G A113067-25G A113067-500G A113067-5G
76687-662EA 35 USD
76687-662 76687-664 76687-666 76687-668 76687-670
Ethyl-3-methyl-4-nitrobenzoate 97%
Ethyl-3-methyl-4-nitrobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₄
MDL Number: MFCD00115804
CAS Number: 30650-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR