(±)-α-Terpineol 98%

Supplier: Ambeed
Warning

Synonyms: α-Terpineol, Terpineol, 2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol

A112949-100G A112949-25G A112949-500G
76687-656EA 16.98 USD
76687-656 76687-658 76687-660
(±)-α-Terpineol 98%
(±)-α-Terpineol

Precaución: for Research Use Only

Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00001557
CAS Number: 98-55-5
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR